Principaux documents
Z708968
ART® non-filtered tip
HLT 10 Reach, size 0.1-10 μL, Pure packed, sterile
Synonyme(s) :
pipet tips, pipette tips
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Volume maximum:
10 μL
Taille:
0.1-10 μL
Stérilité:
sterile
Caractéristiques:
Pure packed
Conditionnement:
pack of 960 ea
Fabricant/nom de marque:
Molecular BioProducts 3511
Matériaux
colorless
Stérilité
sterile
Caractéristiques
Pure packed
Conditionnement
pack of 960 ea
Fabricant/nom de marque
Molecular BioProducts 3511
Volume maximum
10 μL
Taille
0.1-10 μL
Description générale
Extended length for easily reaching the bottom of tubes and a 2 μl reference mark - ideal for micro-volumes.
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.
Informations légales
ART is a registered trademark of Molecular BioProducts, Inc.