Principaux documents
Z659096
Bandelin SONOPULS ultrasonic homogenizer standard and booster horns
SH 70G standard horn for HD 2070
Synonyme(s) :
ultrasonic booster
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.74
Matériaux
titanium
Fabricant/nom de marque
Bandelin
Disponibilité
available only in EU
Diamètre
13 mm
Description générale
Standard and booster horns (Ti-6AI-4V) are furnished with a thread for replaceable probes. With an exterior thread to connect various vessels.