Principaux documents
Z362727
Sigma-Aldrich
Nalgene® polycarbonate Erlenmeyer flask
capacity 125 mL
Synonyme(s) :
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer
About This Item
Code UNSPSC :
41121804
eCl@ss :
32040501
Nomenclature NACRES :
NB.43
Matériaux
flat bottom
polycarbonate
wide-neck
Joint
joint stopper5
Stérilité
non-sterile
Caractéristiques
closure type Stoppers
without cap
Conditionnement
pack of 12
case of 24
Fabricant/nom de marque
Nalgene 4103-0125
Capacité
125 mL
Description générale
Autoclavable; however, repeated autoclaving may cause some loss of mechanical strength.
Autres remarques
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries