Key Documents
GE80-1129-33
GelBond Film 84 X 94 mm, (Pkg/50)
Cytiva 80-1129-33, pack of 50 pieces
About This Item
Code UNSPSC :
41122410
Nomenclature NACRES :
NB.22
Conditionnement
pack of 50 pieces
Fabricant/nom de marque
Cytiva 80-1129-33
Disponibilité
not available in Japan
Température de stockage
room temp
Description générale
GelBond PAGfilm is a polyester film with a treated surface which binds to the agarose gel.
Stockage et stabilité
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Remarque sur l'analyse
To view the Certificate of Analysis for this product, please visit www.cytiva.com.