Principaux documents
EP5490045009
Rotor for Eppendorf® Concentrator Plus
swing-out rotor, A-2-VC, non aerosol tight
About This Item
Code UNSPSC :
41103909
Nomenclature NACRES :
NB.82
Caractéristiques
skirt
to hold 2 x MTP, skirted PCR plates or deepwell plates (max. height 27 mm)
non aerosol tight
swing-out rotor
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 5490045009 (EU)
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models Concentrator Plus
Description générale
Selection of rotors for the Eppendorf Concentrator Plus.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG