Principaux documents
CLS4878
Corning® Transtar® liquid handling system
disposable reservoir liner, 8-channel
Synonyme(s) :
Liquid Handling Equipment
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.11
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
white polystyrene
Stérilité
sterile
Fabricant/nom de marque
Corning 4878
Description générale
The Transtar is a portable, autoclavable liquid handling system designed for use with 96 well plates, A sterile 96 tip disposable cartridge, which loads into the Transtar system, enables liquids to be aspirated, transferred and dispensed over a volume range of 25 - 200 μL in 5 μL increments, Transtar accuracy us rated at 5% at all volume levels.
Informations légales
Corning is a registered trademark of Corning, Inc.
TRANSTAR-96 is a registered trademark of Corning, Inc.