Principaux documents
BAF199210001
Bel-Art® Extended Handle Pestle
cone tip polypropylene, pack of 6 pieces
Synonyme(s) :
grinding and crushing equipment, plastic pestle, tool for manually grinding solid samples
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.74
Matériaux
cone tip polypropylene
Conditionnement
pack of 6 pieces
Fabricant/nom de marque
Bel-Art F19921-0001
L × diam.
21 cm × 4.8 mm
Description générale
Polypropylene pestle for homogenizing in centrifuge, test and grinding tubes
  • Fits conical shaped 1.5 ml tubes
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP