Principaux documents
Z740582
Sigma-Aldrich
Aldrich® Essentials beaker, Griffin
low form, capacity 1000 mL, subdivision, 50 mL (graduations: 50-1000 mL)
About This Item
Code UNSPSC :
41121803
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
Caractéristiques
subdivision 50 mL (graduations: 50-1000 mL)
Conditionnement
pkg of 24 ea
Fabricant/nom de marque
Sigma-Aldrich
Capacité
1000 mL
Description générale
Borosilicate glass, Griffin style, low form beakers with reference graduations and pour spout. Uniform, standard walls and flat bottom for even heating. Meet ASTM E960 designation.
Autres remarques
Browse all glass beakers
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC