Key Documents
Z552720
Sigma-Aldrich
Aldrich® Thermometer Adapter
straight, male joint: ST/NS 29/42, GL 14 thread
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.43
Joint
male joint: ST/NS29/42
Raccords
GL 14 thread
Catégories apparentées
Description générale
Has PTFE faced silicone washer and open-top cap.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC