Principaux documents
Z549681
Sigma-Aldrich
High-vacuum valve adapter
male joint: ST/NS 19/22
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
male joint: ST/NS19/22
Catégories apparentées
Description générale
For high-vacuum applications. Features a high-vacuum PTFE valve. For use with 1/4in. I.D. tubing.