Principaux documents
Z530336
Sigma-Aldrich
Aldrich® reaction vessel bubbler
joint: ST/NS 14/20
Synonyme(s) :
bubblers, scientific bubblers
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS14/20
Capacité
5-7 mL
Catégories apparentées
Description générale
Excellent for closing off a reaction vessel from the atmosphere.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC