Principaux documents
Z309400
Quickfit® flat-bottom boiling flasks
capacity 250 mL, joint: ST/NS 29/32
Synonyme(s) :
FF250/4S
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
flat bottom flask
short
single-neck
Joint
joint: ST/NS29/32
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
case of 5
Fabricant/nom de marque
Quickfit FF 250/4S
Disponibilité
not available in USA and Canada
Capacité
250 mL
Description générale
With short neck and ground-glass socket. Not suitable for vacuum use.
Informations légales
Quickfit is a registered trademark of SciLabware Limited