Principaux documents
Z281883
Sigma-Aldrich
Econo-Press kit
About This Item
Code UNSPSC :
12000000
Nomenclature NACRES :
SB.32
Technique(s)
IR spectroscopy: suitable
Description générale
An inexpensive KBr pellet press to form a 10mm diam. pellet. The Econo-Press includes two bolts and one barrel. Spectra can be obtained with the pellet in the press or the pellet can be placed into an Econo-Card for analysis and storage. The kit contains the Econo-Press, 10 Econo-Cards, and the Econo-Mount holder.