Principaux documents
Z189006
Ace four-neck round-bottom flask
capacity 500 mL
Synonyme(s) :
bottom flask, flask round, flask round bottom
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
four-neck
round bottom flask
Joint
90 degree angle joint ST/NS24/40
center joint ST/NS34/45 vertical
side Joints ST/NS24/40 vertical, except for 500 mL, which is angled (2)
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Ace Glass 6952-04
Capacité
500 mL