Principaux documents
Z180637
Sigma-Aldrich
Ace European-style three-neck flask
capacity 500 mL
Synonyme(s) :
bottom flask, flask round, round flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
flask
round bottom
three-neck
Joint
center joint ST/NS24/40
side joint ST/NS24/40
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Ace Glass 6961-45
Capacité
500 mL
Description générale
Has tapered walls with shallow bottom for more efficient magnetic or mechanical stirring with half-moon shaped paddle stirrer. 7 mm O.D. threaded thermometer port with nylon bushing and FETFE® O-ring.
Informations légales
FETFE is a registered trademark of Ace Glass, Inc.