Principaux documents
Z162159
Spinfin® magnetic stirring bar
H × diam. 9.5 mm × 9.5 mm
Synonyme(s) :
Stir Bar, Stirbar, Stirring Bar
About This Item
Code UNSPSC :
41103806
eCl@ss :
32100100
Nomenclature NACRES :
NB.72
Matériaux
PTFE-coated (fin)
PTFE-coated
Conditionnement
pack of 2
Fabricant/nom de marque
Bel-Art F37125-0038
H × Diam.
9.5 mm × 9.5 mm
Description générale
Disks have fins on top for complete agitation.
Informations légales
Spinfin is a registered trademark of SP Scienceware, a division of SP