Principaux documents
Z103098
Sigma-Aldrich
Aldrich® dry ice condenser
joint: ST/NS 24/40
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS24/40
Description générale
For use with dry ice/acetone slush or liquid nitrogen in cold-finger.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC