Key Documents
WHA10453007
Whatman® pressure filtration devices, MD 050
MD 050/0/18, Connector olive external diam. 9 to 11 mm, for pressure hoses
Synonyme(s) :
Z751383
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.24
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Whatman 10453007
Whatman Article No. 28415707 (US reference)
Caractéristiques et avantages
  • Inlet/outlet connections for MD 050 and MD 142/5 stainless steel pressure filtration devices.
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
Whatman is a registered trademark of Cytiva