Principaux documents
SYNS392942
Synthware cap design stopper
joint: ST/NS 29/42
Synonyme(s) :
Synthware S392942, glass stopper
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS29/42
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S392942
Description générale
For apparatus having an inner opening.
Informations légales
Synthware is a trademark of Kemtech America Inc.