SYNF622425
Synthware™ pressure equalizing addition funnel
25 mL, joint: ST/NS 24/40, stopcock: PTFE
Synonyme(s) :
Synthware F622425, dropping funnel
About This Item
Produits recommandés
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F622425
Capacité
25 mL
25 mL
Description générale
With 1:5 PTFE stopcock
Informations légales
Synthware is a trademark of Kemtech America Inc.