Principaux documents
SYNF612460
Synthware pressure equalizing addition funnel with metering stopcock
60 mL, joint: ST/NS 24/40
Synonyme(s) :
Synthware F612460, dropping funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F612460
Capacité
60 mL
60 mL
Catégories apparentées
Description générale
With PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from full flow to drop wise. The metering plug completely solves the problems of sudden overflow or non-flow during addition of reagents.
Informations légales
Synthware is a trademark of Kemtech America Inc.