Principaux documents
SYNC124400
Synthware inner coil condenser
joint: ST/NS 24/40, hose size 10 mm
Synonyme(s) :
Synthware C124400, Condenser coil, Coil reflux condenser
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware C124400
Taille du tuyau
10 mm
Catégories apparentées
Description générale
Synthwarecoiled condenser features a 400 mm jacketed length for superior cooling efficiency. With standardized 24/40 upper and lower joints, it easily fits various laboratory glassware. The inner coil circulates coolant, maximizing heat exchange. Made from durable borosilicate glass.
Informations légales
Synthware is a trademark of Kemtech America Inc.