Principaux documents
SYNC101100
Synthware Vigreux distillation column
joint: ST/NS 14/20, L 100 mm
Synonyme(s) :
Synthware C101100, distilling column, fractional column
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware C101100
Longueur
100 mm
Catégories apparentées
Description générale
Column length is measured from the highest to lowest indent in the column.
Informations légales
Synthware is a trademark of Kemtech America Inc.