SLW2115/06M
Pyrex® Weighing scoop
10 mL
Synonyme(s) :
Pyrex, SciLabware 2115/06M
About This Item
Produits recommandés
Matériaux
borosilicate glass
Conditionnement
pack of 10 ea
Fabricant/nom de marque
SciLabware 2115/06M
L totale
100 mm
Capacité
10 mL
Description générale
- With hollow stem to aid introduction of the powder into narrow neck vessels e.g. volumetric flasks
- The flat base ensures stability on the balance pan