Key Documents
Z509353
Sigma-Aldrich
Weighing funnel
size L, anti-static PP
Synonyme(s) :
Weighing Apparatus, Weighing Device
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.71
Matériaux
anti-static PP
Taille
L
Conditionnement
pkg of 20 ea
Description générale
  • Sit flat on balance - No rolling, no spilling!
  • Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer
  • Residual sample can be rinsed to guarantee 100% transfer
  • Suitable for safe handling of hazardous materials
  • Available in five different sizes: extra small, small, medium, large, and extra large