Principaux documents
CLS3540
Corning® 384 well microplate, low volume
black polystyrene, round bottom, non-sterile, lid: no, pack of 10
Synonyme(s) :
384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.24
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
black polystyrene
clear bottom
clear wells
round bottom
Stérilité
non-sterile
Caractéristiques
lid: no
skirt
Conditionnement
pack of 10
case of 50
Fabricant/nom de marque
Corning 3540
Taille
384 well
Volume des puits
50 μL
Volume de travail par puits
5-40 μL
Puits
384 wells , low volume
Adéquation
suitable for (molecular, validated for the FLIPR reader)
Type de liaison
non-treated surface
Description générale
  • Corning® 384 well low volume microplates feature a round well design with a clear flat bottom.
  • The non-treated medium binding polystyrene surface is hydrophobic and suitable for the immobilization of antibodies.
  • Ideal for fluorescent homogeneous assays.
Informations légales
Corning is a registered trademark of Corning, Inc.