Key Documents
CAD9920
Animal feeding needle, sterile, disposable, PTFE
size 20G, L × diam. 1.5 in. × 1.9 mm , ball
Synonyme(s) :
PTFE Feeding Needle, Sterile Feeding Needle
About This Item
Code UNSPSC :
42142609
Nomenclature NACRES :
NB.03
Taille
20G
Conditionnement
pkg of 20 x 5 ea (5 needles per sterile Tyvek peel pouch - 20 pouches per box)
L × diam.
1.5 in. × 1.9 mm , ball
Description générale
  • PTFE tube is softer than other tubing types, very flexible, and translucent for easy confirmation of reagent delivery
  • Plastic Luer hubs and soft silicone tips
  • Non-toxic and no-pyrogenic
  • Packaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box