Key Documents
AFM-75009-12
Sigma-Aldrich
12 mm Disc Gripper for AFM
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
NA.23
Description générale
Unique gripper that easily pick up 12 mm diamter AFM/STM discs from a flat surface. Features a protective rubber-coated tip.