Skip to Content
Merck
All Photos(1)

Key Documents

LL24

90073001, human lung, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human lung
growth mode:
Adherent
karyotype:
2n = 46; diploid
morphology:
Fibroblast
products:
Not specified
receptors:
Not specified

Product Name

LL24, 90073001

biological source

human lung

growth mode

Adherent

karyotype

2n = 46; diploid

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human diploid lung

Cell Line Description

Derived from normal tissue taken at post-mortem from the lung of a 5 year old male. An ampoule of cells frozen after one passage and estimated population doubling (PDL) 2 was submitted to the American Type Culture Collection in June, 1977. Derived populations at passage 6 and approximately 14 PDL are available for distribution. Progeny were found to undergo an additional 25 population doublings in longevity trials.

Application

Tumourigenicity studies

Culture Medium

Ham′s F12 + 2mM Glutamine + 15% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Finite lifespan, a further 20 population doublings expected.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service