Skip to Content
Merck
All Photos(1)

Key Documents

IEC 6

88071401, rat intestine, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

IEC 6, 88071401

biological source

rat intestine

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat small intestine epithelial

Cell Line Description

Normal rat small intestine epithelial cells which synthesise fibronectin and collagen. Growth is inhibited by cortisol. Cells possess cell surface antigens specific for intestinal epithelial cells in vivo. Capable of at least 10 population doublings.

Application

Small bowel biology

Culture Medium

DMEM + 2mM Glutamine + 0.1 IU/ml Insulin + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service