Skip to Content
Merck
All Photos(1)

Key Documents

Mero-95

9100108, human lung, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Mero-95, 09100108

biological source

human lung

growth mode

Adherent

karyotype

Number of chromosomes 54-58

morphology

Epithelial

products

Epithelial membrane antigen (EMA)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

A human mesothelioma cell line derived from a malignant mesothelioma. Asbestos exposure

Cell Line Description

Mero-95 was derived from a pleural effusion

Application

Investigation of the properties of malignant mesothelioma cells and the development of new diagnostic methods

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 11,12
D16S539: 11
D5S818: 11,14
D7S820: 7,9
THO1: 6
TPOX: 8,12
vWA: 18,19

Culture Medium

Hams F10 + 15% FCS + 2mM Glutamine

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Population doubling approx 30 hours

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service