product name
Mero-82, 09100105
biological source
human lung
growth mode
Adherent
karyotype
Number of chromosomes 48-50/94-98
morphology
Epithelial
products
Epithelial membrane antigen (EMA)
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
A human mesothelioma cell line derived from a malignant mesothelioma. Asbestos exposure
Cell Line Description
Mero-82 was derived from a pleural effusion
Application
Investigation of the properties of malignant mesothelioma cells and the development of new diagnostic methods
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 10,13
D13S317: 12
D16S539: 12
D5S818: 11,12
D7S820: 10
THO1: 9.3
TPOX: 8
vWA: 16
CSF1PO: 10,13
D13S317: 12
D16S539: 12
D5S818: 11,12
D7S820: 10
THO1: 9.3
TPOX: 8
vWA: 16
Culture Medium
Hams F10 + 15% FCS + 2mM Glutamine
Subculture Routine
Split sub-confluent cultures (70-80%) 1:4 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Population doubling approx 24 hours
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service