Skip to Content
Merck
All Photos(1)

Key Documents

MRC-5 pd25 Cell Line human

5081101, human lung, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MRC-5 pd25 Cell Line human, fetal lung, 05081101

biological source

human lung

growth mode

Adherent

karyotype

2n=46, diploid

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human foetal lung

Cell Line Description

Established from normal lung tissue of a 14 week old male foetus. The cells undergo between 60-70 population doublings before senescence. The virus susceptibility of this line to human viruses is similar to WI-38.

Application

Established from normal lung tissue of a 14 week old male foetus. The cells undergo between 60-70 population doublings before senescence. The virus susceptibility of this line to human viruses is similar to WI-38.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 11,14
D16S539: 9,11
D5S818: 11,12
D7S820: 10,11
THO1: 8
TPOX: 8
vWA: 15

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service