SYNPLE-BT013
Reagent-cartridge Biotin tag-PEG3-O-4-P-NH
Synple
Synonym(s):
Synple 2
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
MDL number:
UNSPSC Code:
12161503
Quality Level
parameter
up to 0.1 mmol scale
Looking for similar products? Visit Product Comparison Guide
General description
Reagent-cartridge Biotin tag-PEG3- O-4-P-NH is a reagent cartridge for the Synple 2 Automated Synthesizer that enables the biotin tagging of aldehydes and ketones through a reductive amination. The cartridge contains all necessary reagents for linking the chosen aldehyde or ketone substrate to biotin.
Scale: up to 0.1 mmol
Linker: PEG3
Terminal functional group: secondary amine
Compatible substrates: aldehyde, ketone
Cartridge contents:
Related Cartridges:
Learn more about the Synple automated synthesis platform.
Features and Benefits of Synple Platform
Scale: up to 0.1 mmol
Linker: PEG3
Terminal functional group: secondary amine
Compatible substrates: aldehyde, ketone
Cartridge contents:
- Reducing agent (supported cyanoborohydride)
- Base (supported triethylamine)
- SCX (for purification)
- Biotin linker
Related Cartridges:
- SYNPLE-BT002Reagent-cartridge Biotin tag-PEG2-NH2
- SYNPLE-BT003Reagent-cartridge Biotin tag-PEG3-NH2
- SYNPLE-BT004Reagent-cartridge Biotin tag-PEG4-NH2
- SYNPLE-BT012Reagent-cartridge Biotin tag-PEG2-O-4-P-NH
- SYNPLE-BT014Reagent-cartridge Biotin tag-PEG4-O-4-P-NH
- SYNPLE-BT022Reagent-cartridge Biotin tag-PEG2-a-COOTfp
- SYNPLE-BT023Reagent-cartridge Biotin tag-PEG3-a-COOTfp
- SYNPLE-BT024Reagent-cartridge Biotin tag-PEG4-a-COOTfp
- SYNPLE-BT032Reagent-cartridge Biotin tag-PEG2-COOTfp
- SYNPLE-BT033Reagent-cartridge Biotin tag-PEG3-COOTfp
- SYNPLE-BT034Reagent-cartridge Biotin tag-PEG4-COOTfp
Learn more about the Synple automated synthesis platform.
Features and Benefits of Synple Platform
- Time savings - pre-installed protocols have been highly optimized, which obviates the need for the user′s own reaction development and optimization
- Easy to use - protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
- Safety - all chemicals are contained in the system, minimizing the user′s exposure
- Set-up reaction in less than 5 minutes
- Small footprint
- Plug & Play system - simple installation
- Reaction temperatures from 25 °C - 80 °C
- Cartridges available for many different reactions
- Prefilled with all required reagents
- RFID chip containing optimized reaction protocol
Legal Information
Product of Synple Chem AG
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service