SYNPLE-P025
Reagent-cartridge VHL-PEG5
Synple
Synonym(s):
Synple 2
About This Item
Recommended Products
Looking for similar products? Visit Product Comparison Guide
General description
Scale: up to 0.1 mmol
Linker: PEG5
E3 ligase target and ligand: von Hippel-Lindau (VHL), (S,R,S)-AHPC
Terminal functional group: amine
Compatible substrates: ketone, aldehyde
Cartridge contents:
- Reducing agent (supported cyanoborohydride)
- Base (supported triethylamine)
- SCX (for purification)
- Degrader building block linker reagent
Related Cartridges:
- SYNPLE-P001Reagent-cartridge CRBN-PEG1
- SYNPLE-P002Reagent-cartridge CRBN-PEG2
- SYNPLE-P003Reagent-cartridge CRBN-PEG3
- SYNPLE-P004Reagent-cartridge CRBN-PEG4
- SYNPLE-P005Reagent-cartridge CRBN-PEG5
- SYNPLE-P021Reagent-cartridge VHL-PEG1
- SYNPLE-P022Reagent-cartridge VHL-PEG2
- SYNPLE-P023Reagent-cartridge VHL-PEG3
- SYNPLE-P024Reagent-cartridge VHL-PEG4
Learn more about the Synple automated synthesis platform.
Learn more about protein degradation.
Explore all our Protein Degrader Building Blocks
Features and Benefits of Synple Platform
- Time savings – pre-installed protocols have been highly optimized, which obviates the need for the user′s own reaction development and optimization
- Easy to use – protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
- Safety – all chemicals are contained in the system, minimizing the user′s exposure
- Set-up reaction in less than 5 minutes
- Small footprint
- Plug & Play system – simple installation
- Reaction temperatures from 25 °C - 80 °C
- Cartridges available for many different reactions
- Prefilled with all required reagents
- RFID chip containing optimized reaction protocol
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.
Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.
Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.
Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service