Skip to Content
Merck
All Photos(1)

Documents

80490

Sigma-Aldrich

Dimethyl pimelimidate dihydrochloride

≥99.0% (AT)

Synonym(s):

DMP, Dimethyl pimelinediimidate dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3OC(=NH)CH2CH2]2CH2 · 2HCl
CAS Number:
Molecular Weight:
259.17
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99.0% (AT)

form

powder

storage temp.

2-8°C

SMILES string

Cl.Cl.COC(=N)CCCCCC(=N)OC

InChI

1S/C9H18N2O2.2ClH/c1-12-8(10)6-4-3-5-7-9(11)13-2;;/h10-11H,3-7H2,1-2H3;2*1H

InChI key

LRHXBHUTQWIZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Cross-linking reagent for the modification of proteins via amidation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of a diimidoester cross-linking reagent to examine the subunit structure of rabbit muscle pyruvate kinase.
G E Davies et al.
Canadian journal of biochemistry, 50(4), 416-422 (1972-04-01)
C Schneider et al.
The Journal of biological chemistry, 257(18), 10766-10769 (1982-09-25)
A method is described by which an immunoaffinity matrix was constructed by binding antibody directly or indirectly to protein A-Sepharose 4B followed by cross-linking of the complex with dimethyl pimelimidate. This allows optimal spatial orientation of antibodies and, thus, maximum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service