Skip to Content
Merck
Dimethyl pimelimidate dihydrochloride ≥99.0% (AT)

Dimethyl pimelimidate dihydrochloride

Dimethyl pimelimidate dihydrochloride ≥99.0% (AT)
Synonym(s):
DMP, Dimethyl pimelinediimidate dihydrochloride
Linear Formula:
[CH3OC(=NH)CH2CH2]2CH2 · 2HCl
CAS Number:
Molecular Weight:
259.17
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing