Documents
57270
Glass Magnet
diam. × thickness 4 in. × 3/16 in., pkg of 2 ea
Synonyme(s) :
Glass Magnet FLACON HOLDER boîte de 2
About This Item
Code UNSPSC :
41122001
Conditionnement
pkg of 2 ea
Diam. × épaisseur
4 in. × 3/16 in.
Description générale
The Glass Magnet Vial Holder keeps individual vials, ampuls, and other vessels in place.
Informations légales
Glass Magnet is a trademark of Sigma-Aldrich Co. LLC
Produit(s) apparenté(s)
Choose from one of the most recent versions:
Certificats d'analyse (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Service Clients
Déjà en possession de ce produit ?
Articles
Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.