Documents
57101-U
Supelco
Système d′extraction sous vide Visiprep pour 5 fioles
DL (Disposable Liner)
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.21
Technique(s)
solid phase extraction (SPE): suitable
Application(s)
food and beverages
Description générale
Contrairement au modèle Visisprep standard, le modèle Visiprep DL (liner jetable) évite les contaminations croisées lorsque des échantillons sont extraits successivement sur le même port. Les liners en PTFE jetables sont passés à travers chaque port du système DL et permettent le passage du solvant directement à travers l′aiguille-guide jusque dans la fiole de collection (fiole de 50mL) Les réactifs et échantillons sortant du tube SPE n′étant en contact qu′avec la surface interne du liner jetable, le risque de contamination croisée lors de la préparation d′échantillons multiples, est virtuellement éliminé.
The Visiprep 5-Port Flask Vacuum Manifold enables analysts using Supelco solid phase extraction tubes to simultaneously prepare up to 5 samples. Unlike conventional vacuum manifolds, the Visiprep 5-Port Flask Manifold allows users to collect their SPE eluate directly into 50 mL round or flat bottom flasks for direct Rotovap evaporation. The manifold consists of a chemical resistant 5-port cover (DL or standard available), gasket, base, a glass basin, vacuum gauge and bleed valve, 5 flow control valves, 5 replaceable solvent guide needles, and a base plate that supports up to five 50 mL round or flat bottom flasks.
Each port on both the standard and DL Visiprep models are equipped with flow control valves.
Recommended Flasks: Aldrich single-neck flask, 50 mL, joint: ST/NS 24/40
  • Round Bottom (Cat. No. Z414484)
  • Flat Bottom (Cat. No. Z418773)
Caractéristiques et avantages
Supplied with 100 disposable liners (Part No. 57059).
Informations légales
3D Insert is a trademark of 3D Biotek, LLC
Visiprep is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificats d'analyse (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Service Clients
Déjà en possession de ce produit ?
Chun-Lin Fan et al.
Journal of AOAC International, 98(1), 130-148 (2015-04-11)
A 3 month study was conducted on the ruggedness of a multiresidue method for accuracy and stability. The results indicate that in terms of Youden pair ratios of 201 pesticide aged tea samples falling approximately within 1.00-1.20 of the ratio