Principaux documents
24867
Supelco
Support de tube pour pompe PAS-500
for use with 6mm sorbent tube
About This Item
Code UNSPSC :
41104008
Technique(s)
active air sampling: suitable
Application(s)
air monitoring
environmental
industrial hygiene
Compatibilité
for use with 6mm sorbent tube
Description générale
Tube Holder for PAS-500 Pump for 6mm sorbent tubes is used as accessories for PAS-500 micro air sampler used in air monitoring.