Principaux documents
Z719560
Sorenson low binding E tips
maximum volume 10 μL, case of 4800 ea (50 racks of 96 tips), sterile
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Matériaux
colorless
polypropylene
Stérilité
sterile
Caractéristiques
barrier: no
Conditionnement
case of 4800 ea (50 racks of 96 tips)
pack of 960 ea (10 racks of 96 tips)
Fabricant/nom de marque
Sorenson 22210T
Volume maximum
10 μL
Volume
10 μL
Description générale
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Extended length. Fits Eppendorf ultra-micro pipettors.
Informations légales
Sorenson is a trademark of Sorenson BioScience, Inc.