Documents
Z694673
3D Biotek Polycaprolactone (PCL) scaffold inserts
for 3D bioreactor 24 well chamber, fiber diam. ~300 μm
Synonyme(s) :
3D, 3D Cell Culture
About This Item
Code UNSPSC :
41122107
Nomenclature NACRES :
NB.22
Stérilité
sterile
Fabricant/nom de marque
3D Biotek PCL303024-BR
Diam. des fibres
~300 μm
Dimension de pores
300 μm (Fiber spacing)
Type de liaison
non-treated surface