Principaux documents
Z376221
Sigma-Aldrich
PCR Workstation
W × D × H 48 in. × 24 in. × 24 in., AC/DC input 240 V AC, Euro plug
About This Item
Code UNSPSC :
41106300
Nomenclature NACRES :
NB.22
Technique(s)
PCR: suitable
Larg. × P × H
48 in. × 24 in. × 24 in.
Description générale
A self-contained work area that will help protect your PCR runs against contamination. Dual UV bulbs irradiate the work area prior to use, reducing the possibility that contaminating DNA will be amplified. Containment features reduce the chance of airborne contamination.
The non-ventilated hood has acrylic windows for a clear view, an acid-resistant black formica work surface, a laminated safety glass fascia, and double access doors. The germicidal lamps are controlled by a 24-hr timer. The work light is provided by dual fluorescent lamps, and there is a ceiling-mounted double AC outlet. Replacement part sold separately.
Informations légales
PCR Workstation is a trademark of C.B.S. Scientific Co.