Principaux documents
Z370371
Sigma-Aldrich
Carestream® BioMax® light film
Synonyme(s) :
autoradiography film, xray film
About This Item
Code UNSPSC :
41105341
Nomenclature NACRES :
NB.22
Taille
8 in. (20 cm) × 10 in. (25 cm)
Description générale
Double-emulsion film on clear base. Provides clearest background and highest sensitivity for chemiluminescence detection systems. Also suitable for autoradiography, but with less sensitivity for high-energy isotopes than X-Omat AR and BioMax MS, and than BioMax MR for medium-energy isotopes.
Sheets of film with no interleaved paper
Informations légales
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Manav Gupta et al.
Cancer research, 80(18), 3841-3854 (2020-07-22)
Inactivation of SMARCA4/BRG1, the core ATPase subunit of mammalian SWI/SNF complexes, occurs at very high frequencies in non-small cell lung cancers (NSCLC). There are no targeted therapies for this subset of lung cancers, nor is it known how mutations in
Ana M Hernández-Vega et al.
Cells, 9(9) (2020-08-23)
The mesenchymal phenotype of glioblastoma multiforme (GBM), the most frequent and malignant brain tumor, is associated with the worst prognosis. The epithelial-mesenchymal transition (EMT) is a cell plasticity mechanism involved in GBM malignancy. In this study, we determined 17β-estradiol (E2)-induced
Shaista Naqvi et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(19), 7762-7767 (2009-05-07)
Vitamin deficiency affects up to 50% of the world's population, disproportionately impacting on developing countries where populations endure monotonous, cereal-rich diets. Transgenic plants offer an effective way to increase the vitamin content of staple crops, but thus far it has
Steven C Chen et al.
PloS one, 6(5), e19780-e19780 (2011-05-24)
Although recent publications have linked the molecular events driving facioscapulohumeral muscular dystrophy (FSHD) to expression of the double homeobox transcription factor DUX4, overexpression of FRG1 has been proposed as one alternative causal agent as mice overexpressing FRG1 present with muscular
Articles
Background and protocols describing the various methods used by molecular biologists to detect samples of protein or nucleic acids bound to membranes.
Questions
1–2 of 2 Questions  
  1. Products Z370371 and Z370401 are both described as Carestream® Kodak® BioMax® light film size 8 in. (20 cm) × 10 in. (25 cm). Both are listed as 50 sheet per order. What is the difference between the 2 products?
    1 answer
    1. Product Z370371 has a Carestream Health Inc product number of 178 8207.  The Carestream website indicates the sheets are non-interleaved (not individually wrapped)  for product 178 8207.  Product Z370401 has a Carestream product number of 876-1520.  This product has individual wrapping for each sheet of film.
      Helpful?
  2. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.