Principaux documents
Z350370
Sigma-Aldrich
Carestream® BioMax® MR film
Synonyme(s) :
autoradiography film, xray film
About This Item
Code UNSPSC :
41105341
Nomenclature NACRES :
NB.22
Taille
8 in. (20 cm) × 10 in. (25 cm)
Description générale
Sheets of film with no interleaved paper
Single-emulsion film that provides maximum resolution and clearest background. The most sensitive film for direct 14C, 35S, and 33P autoradiography. Higher resolution than X-OMAT AR and BioMax MS for 32P direct and screen exposures and chemiluminescence, but with reduced sensitivity.
Informations légales
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Certificats d'analyse (COA)
Lot/Batch Number
Désolés, nous n'avons pas de COA pour ce produit disponible en ligne pour le moment.
Shujuan Gao et al.
Journal of molecular signaling, 6, 8-8 (2011-08-13)
The family of A-kinase-anchoring proteins, AKAPs, constitutes a group of molecular scaffolds that act to catalyze dynamic interactions of protein kinase A, protein kinase C, tyrosine kinases, G-protein-coupled receptors and ion channels. AKAP5 (MW ~47 kDa) and AKAP12 (MW ~191
Atsuko Kimura et al.
The European journal of neuroscience, 30(2), 299-306 (2009-07-21)
The 5-HT(2C) receptor has been implicated in mood and eating disorders. In general, it is accepted that 5-HT(2C) receptor agonists increase anxiety behaviours and induce hypophagia. However, pharmacological analysis of the roles of these receptors is hampered by the lack
Articles
Background and protocols describing the various methods used by molecular biologists to detect samples of protein or nucleic acids bound to membranes.