Principaux documents
Z330000
BRAND® narrow-mouth bottle, LDPE
capacity 2000 mL
Synonyme(s) :
ldpe bottle, polyethylene bottles, polyethylene containers
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Matériaux
low-density polyethylene bottle
narrow-mouth bottle
round bottle
translucent
Joint
Joints threaded neck (GL28)
Description
dropper nozzle caps available
Caractéristiques
with cap
Conditionnement
pkg of 25
Fabricant/nom de marque
BRAND 129464
Capacité
2000 mL
Diam. × H
117 mm × 264 mm
Description générale
Sparsame Vielzweck Flasche
Autres remarques
BR129464 (India)
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG