Principaux documents
WHAWB120206
QIAcard Indicating FTA Cards
classic, 4 sample areas per card, pkg of 100 cards
Synonyme(s) :
QIAcard FTA Indicating Classic (100), Whatman FTA, Z719749, whatman fta cards
1 PKG
1 040,00 $
1 040,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 PKG
1 040,00 $
About This Item
Code UNSPSC :
41105500
Nomenclature NACRES :
NB.22
1 040,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Description
indicating FTA classic card with 4 sample areas per card, 125 μL maximum volume/sample area, 500 μL maximum total volume/card
Fabricant/nom de marque
Qiagen WB120206
Conditionnement
pkg of 100 cards
Description générale
FTA Cards offer a compact room-temperature storage system that reduces the need for precious freezer space.
QIAcard FTA classic format with 4 spot area to collect, stabilize, process, transport, and archive colored samples such as blood. FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
FTA classic cards utilize QIAcard FTA technology that simplifies the handling and processing of nucleic acids. FTA Cards contain chemicals that lyse cells, denature proteins and protect nucleic acids from nucleases, oxidation and UV damage. FTA cards rapidly inactivate organisms, including blood-borne pathogens, and prevent the growth of bacteria and other microorganisms.
Convenient for protocols that require different locations for testing and archiving samples. Different samples can be processed independently.
Caractéristiques et avantages
  • Includes a pink dye that turns white when a colorless sample is applied.
  • 4 sample areas for application of up to 500 μl sample volume per card.
  • Simply apply your sample to the FTA Card. Cell membranes and organelles are lysed and the released nucleic acids are entrapped in the fibers of the matrix. The nucleic acids remain immobilized and are preserved for transport, immediate processing or long-term room temperature storage.
  • Since captured nucleic acids are preserved, FTA Cards facilitate sample collection in remote locations and simplify sample transport.
  • Ship your samples back to the laboratory without expensive special handling or dry ice and process at your convenience.
  • FTA Cards can be used with virtually any sample type: Blood, Cultured cells, Buccal cells, Plasmids, and Solid tissue.
  • Captured nucleic acids are ready for purification when you are. Just take a punch from the FTA Card, wash with FTA Purification Reagent and rinse with TE-1 (10 mM Tris-HCI, 0.1 mM EDTA, pH 8) buffer. DNA on the washed punch is ready to use in applications such as PCR, SNP analysis, and real-time PCR. Since PCR products remain in solution, the punch can be used for multiple amplifications.
Autres remarques
Sample integrity is optimized when FTA Cards are stored in a Multi-Barrier Pouch with a Desiccant Packet.
Field of Use : For internal research use only
Informations légales
FTA is a trademark of Qiagen Group
QIAcard is a trademark of Qiagen Group
Articles
Description of common problems that can occur during genomic DNA preparation
This article describes the evaluation of Whatman FTA cards from Cytiva for their ability to collect, store, and isolate high-quality RNA from a variety of crude biological samples.
Polymerase chain reaction (PCR) is a technique that results in exponential amplification of a target DNA sequence.
Protocoles
This page shows genomic DNA purification from sample on FTA elute from Cytiva.
This document shows intructions for EasiCollect™ device from Cytiva.
This study examined the yield and quality of DNA from samples applied to Whatman FTA cards, using five common methods of DNA extraction.
A number of options are available for sample collection, transport, archiving, and DNA purification.
Afficher tout