Principaux documents
WHAWB100014
QIAcard Bloodstain non-indicating FTA cards
pack of 100 cards
Synonyme(s) :
QIAcard Bloodstain (100), Whatman FTA, Z719838, blood stain cards
1 PKG
359,00 $
359,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 PKG
359,00 $
About This Item
Code UNSPSC :
41105500
Nomenclature NACRES :
NB.22
359,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Fabricant/nom de marque
Qiagen WB100014
Conditionnement
pack of 100 cards
Adéquation
suitable for (collection and transport of blood and bodily fluids)
Description générale
The QIAcard Bloodstain (4 sample areas per card) is a non-Indicating and non-FTA treated card designed for the collection and short-term handling of blood and bodily fluids. Unlike FTA Cards, the Bloodstain Cards are not coated with a protective chemical cocktail and DNA is as risk of degradation and damage. Convenient transport of blood and bodily fluids with protective cover included.
Caractéristiques et avantages
  • Designed for the collection and transport of blood and bodily fluids
  • Each card features a protective cover
  • Blood stain cards are designed for short-term handling of specimens
  • Card enables efficient sample collection
  • 125 μL sample per spot
Autres remarques
Field of Use : For internal research use only
Informations légales
FTA is a trademark of Qiagen Group
QIAcard is a trademark of Qiagen Group
Protocoles
This document shows intructions for EasiCollect™ device from Cytiva.