Principaux documents
TTRT96P250W-EP
Eppendorf twin.tec® Real-Time PCR Plates 96
Semi-Skirted, PCR clean, volume 250 μL, white plate, pack of 25
About This Item
Code UNSPSC :
41106300
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
polypropylene wells
white plate
Caractéristiques
skirt
PCR clean
Semi-Skirted
Conditionnement
pack of 25
Fabricant/nom de marque
Eppendorf® 951022055 (NA Only)
Technique(s)
PCR: suitable
Taille
96 well
Volume
250 μL
Volume de travail
250 μL
Description générale
The white wells of Eppendorf’s twin.tec real-time PCR plates increase the reflection of fluorescent light in real-time PCR (qPCR) and other fluorescence assays. The increased fluorescence from these specialized qPCR plates improves the sensitivity of your assays, while also improving reproducibility and consistency of your results.
Caractéristiques et avantages
  • White wells for better reflection
  • High mechanical stability
  • Raised rims for effective sealing
  • Skirted and semi-skirted frames
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121 °C, 20 min)
  • PCR clean and Eppendorf Forensic DNA Grade s available
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG