Principaux documents
TMO312085-0002
Nalgene® narrow-mouth bottle
amber high-density polyethylene, size 60 mL, case of 1000 ea
Synonyme(s) :
Nalgene bottle, plastic bottle
1000 EACH
1 120,00 $
1 120,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1000 EACH
1 120,00 $
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
1 120,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
amber high-density polyethylene
Conditionnement
case of 1000 ea
Fabricant/nom de marque
Nalgene 312085-0002
Taille
60 mL
Description générale
Store and ship light-sensitive reagents with Thermo Scientific Nalgene Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure. Ideal for diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals, these high density polyethylene bottles are injection blow-molded for uniform wall thickness and strength. They meet USP light transmission standards and are bulk packed for packaging applications.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries